Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEMA7A Monoclonal Antibody (1D5G8)

Supplier: Thermo Scientific MA538466
Description
Specifications
| SEMA7A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O75326 | |
| SEMA7A | |
| Purified recombinant fragment of human CD108 (aa: 48-222) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 1D5G8 | |
| Unconjugated | |
| SEMA7A | |
| 2900057C09Rik; CD108; CDw108; H-Sema K1; H-SEMA-K1; H-Sema-L; JMH; JMH blood group antigen; John Milton Hagen blood group H-Sema K1; john-Milton-Hargen human blood group Ag; MGC126692; MGC126696; M-Sema-L; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7A; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7A (JMH blood group); sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, 7A; sema K1; Sema L; SEMA7A; SEMAK1; SEMAL; semaphorin 7A (John Milton Hagen blood group); semaphorin 7A, GPI membrane anchor; semaphorin 7A, GPI membrane anchor (John Milton Hagen blood group); Semaphorin K1; semaphorin-7A; Semaphorin-K1; semaphorin-L; Semk1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 8482 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction