Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEMA7A Monoclonal Antibody (MEM-150), FITC
GREENER_CHOICE

Catalog No. 01676319
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-319 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-319 Supplier Invitrogen™ Supplier No. MA119589
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with CD108 (JMH blood group antigen), a 80 kDa GPI-anchored extracellular glycoprotein expressed on various cell types including erythrocytes, lymphoblasts; at low levels it is present on circulating lymphocytes.

CD108 (Sema7A) is a GPI-anchored semaphorin family member, which enhances central and peripheral axonal growth and is required for proper axon track formation during ebryogenesis. CD108 also regulates osteoclast differentiation and pre-osteoblastic cell migration, and in immune system affects cell proliferation, chemotaxis and cytokine release. On erythrocytes CD108 defines the JMH (John-Milton-Hagen) human blood group. CD108 signalizes through its receptors - plexin C1 and beta1 integrins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMA7A
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-150
Conjugate FITC
Formulation TBS with 0.2% BSA and 15mM sodium azide; pH 8.0
Gene SEMA7A
Gene Accession No. O75326
Gene Alias 2900057C09Rik; CD108; CDw108; H-Sema K1; H-SEMA-K1; H-Sema-L; JMH; JMH blood group antigen; John Milton Hagen blood group H-Sema K1; john-Milton-Hargen human blood group Ag; MGC126692; MGC126696; M-Sema-L; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7A; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7A (JMH blood group); sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, 7A; sema K1; Sema L; SEMA7A; SEMAK1; SEMAL; semaphorin 7A (John Milton Hagen blood group); semaphorin 7A, GPI membrane anchor; semaphorin 7A, GPI membrane anchor (John Milton Hagen blood group); Semaphorin K1; semaphorin-7A; Semaphorin-K1; semaphorin-L; Semk1
Gene Symbols SEMA7A
Host Species Mouse
Immunogen T cell line HPB-ALL.
Purification Method Size-exclusion chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8482
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.