Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEMA7A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 180701AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-1969


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

CD108 (Sema7A) is a GPI-anchored semaphorin family member, which enhances central and peripheral axonal growth and is required for proper axon track formation during embryogenesis. CD108 also regulates osteoclast differentiation and pre-osteoblastic cell migration, and in immune system affects cell proliferation, chemotaxis and cytokine release. On erythrocytes CD108 defines the JMH (John-Milton-Hagen) human blood group. CD108 signalizes through its receptors - plexin C1 and beta1 integrins.
Specifications

Specifications

SEMA7A
Polyclonal
Unconjugated
SEMA7A
CD108, CDw108, H SEMA K1, H Sema L, JMH, JMH blood group antigen, Sema K1, Sema L, SEMA7A, SEMAK1, SEMAL, Semaphorin 7A, Semaphorin K1, Semaphorin L
Rabbit
Antigen Affinity Chromatography
RUO
20361, 8482
-20°C
Liquid
Immunoprecipitation, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O75326, Q9QUR8
SEMA7A
SEMA7A Fusion Protein Ag12707
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.