Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Semaphorin 3A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323223
Description
Semaphorin 3A Polyclonal antibody specifically detects Semaphorin 3A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
Semaphorin 3A | |
Polyclonal | |
Western Blot 1:1000, Immunohistochemistry 6:00, Immunocytochemistry/Immunofluorescence 1:100 | |
(semaphorin) 3A, coll-1, collapsin 1, Hsema-I, Hsema-III, MGC133243, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema III, SEMA1, SEMAD, SEMAIII, SEMAL, semaphorin D, Semaphorin III, semaphorin L, semaphorin-3A, SemD | |
Semaphorin 3A synthetic peptide (coupled to carrier protein) corresponding to amino acids in the central region of human Semaphorin 3A. The sequence used is highly conserved in rat and mouse Semaphorin 3A, and has low homology to other semaphorin family members. [UniProt# Q14563] | |
0.1 mL | |
Neuroscience | |
10371 | |
Store at -20°C. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction