Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Semaphorin 3E Antibody (400513) [DyLight 405], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB32391E
Description
Semaphorin 3E Monoclonal antibody specifically detects Semaphorin 3E in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOFSpecifications
Semaphorin 3E | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), CyTOF | |
400513 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
(semaphorin) 3E, coll-5, KIAA0331M-SEMAH, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, SEMAHM-SemaK, semaphorin-3E | |
Mouse myeloma cell line NS0-derived recombinant human Semaphorin 3E, Thr25-Ser775 (Arg557Ala and Arg560Ala), Accession # O15041 | |
0.1 mL | |
Neuroscience | |
9723 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction