Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Semaphorin 3F Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323386
Description
Semaphorin 3F Polyclonal antibody specifically detects Semaphorin 3F in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Semaphorin 3F | |
Polyclonal | |
Western Blot 1:1000 | |
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3F, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, 3F, sema III/F, sema IV, SEMA4, SEMA-IV, semaphorin III/F, semaphorin IV | |
Semaphorin 3F synthetic peptide (coupled to carrier protein) corresponds to amino acids from the C-terminal region of human Sema-3F. This sequence is highly conserved in rat and mouse Sema-3F, and has low homology to other semaphorin family members. [UniProt# Q13275] | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
6405 | |
Store at -20°C. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction