Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Semaphorin 4D/CD100 Antibody (006), DyLight 405

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290665V
Description
Semaphorin 4D/CD100 Monoclonal specifically detects Semaphorin 4D/CD100 in Mouse samples. It is validated for Sandwich ELISA Detection.Specifications
| Semaphorin 4D/CD100 | |
| Monoclonal | |
| DyLight 405 | |
| A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Semaphorin 4D/CD100 (NP_038688.2; Met1-Arg733). | |
| 0.1 mL | |
| Apoptosis, Immunology | |
| 10507 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 6 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction