Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Semaphorin 4D/CD100 Antibody (010), Alexa Fluor™ 488

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290666AF488
Description
Semaphorin 4D/CD100 Monoclonal specifically detects Semaphorin 4D/CD100 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Semaphorin 4D/CD100 | |
Monoclonal | |
Alexa Fluor 488 | |
A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Semaphorin 4D/CD100 (NP_038688.2; Met1-Arg733). | |
0.1 mL | |
Apoptosis, Immunology | |
10507 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, Immunofluorescence | |
10 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction