Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Semaphorin 4D/CD100 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00
Specifications
Antigen | Semaphorin 4D/CD100 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017915
![]() |
Novus Biologicals
NB10093562SS |
0.025 mg |
Each for $206.00
|
|
|||||
NB10093562
![]() |
Novus Biologicals
NB10093562 |
0.1 mg | N/A | N/A | N/A | ||||
Description
Semaphorin 4D/CD100 Polyclonal specifically detects Semaphorin 4D/CD100 in Human samples. It is validated for Western Blot.Specifications
Semaphorin 4D/CD100 | |
Western Blot 1:100-1:2000 | |
Polyclonal | |
Rabbit | |
Apoptosis, Immunology | |
PBS with 0.02% Sodium Azide | |
10507 | |
Synthetic peptide corresponding to residues 777-788 [KPKSDFCDREQS] of the human CD100 protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D | |
SEMA4D | |
IgG | |
Affinity Purified | |
CD100 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title