Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Semaphorin 4D/CD100 Mouse, Clone: SEMA4D/933, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | Semaphorin 4D/CD100 |
---|---|
Clone | SEMA4D/933 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready |
Applications | Flow Cytometry, Immunofluorescence, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP247796B
![]() |
Novus Biologicals
NBP2477960.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Semaphorin 4D/CD100 Monoclonal specifically detects Semaphorin 4D/CD100 in Human, Primate samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, CyTOF-ready.Specifications
Semaphorin 4D/CD100 | |
1.0 mg/mL | |
Flow Cytometry, Immunofluorescence, CyTOF | |
Unconjugated | |
Mouse | |
Apoptosis, Immunology | |
PBS with No Preservative | |
10507 | |
Human recombinant SEMA4D protein | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
SEMA4D/933 | |
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Primate | |
A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D | |
SEMA4D | |
IgG1 κ | |
Protein A or G purified | |
Recognizes a homodimeric protein comprised of 50kDa subunits, identified as CD100. It is expressed on majority of haemopoietic cells (B, T, NK and myeloid cells) and is absent from bone marrow, erythrocytes, eosinophils and endothelial cells. Its expression is increased after PHA-activation. CD100 was shown to associate with different partner molecules in T cells such as CD45, a key molecule with protein tyrosine phosphatase activity involved in T-cell transduction, and a Serine kinase. It plays a role in homotypic cell adhesion and in T cell activation. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title