Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Semaphorin 4F Antibody (780225), Biotin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB7200B
Description
Semaphorin 4F Monoclonal antibody specifically detects Semaphorin 4F in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
Semaphorin 4F | |
Monoclonal | |
Biotin | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
780225 | |
Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
M-SEMA, m-Sema-M, PRO2353, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4F, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4F, Sema M, Sema W, SEMAM, semaphorin-4F, semaphorin-M, Semaphorin-W, SEMAWsemaphorin M | |
Mouse myeloma cell line NS0-derived recombinant mouse Sema-4F, Arg41-Gly667, Accession # Q9Z123 | |
0.1 mL | |
Neuroscience | |
10505 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction