Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Semaphorin 4G Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297839UV
Description
Semaphorin 4G Polyclonal specifically detects Semaphorin 4G in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Semaphorin 4G | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
KIAA1619FLJ20590, MGC102867, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4G, semaphorin-4G | |
Produced in rabbits immunized with E. coli-derived Human Semaphorin 4G fragment. | |
0.1 mL | |
Neuroscience | |
57715 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction