Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Semaphorin 5A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | Semaphorin 5A |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Semaphorin 5A Polyclonal specifically detects Semaphorin 5A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Semaphorin 5A | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q13591 | |
9037 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:PWTKCSATCGGGHYMRTRSCSNPAPAYGGDICLGLHTEEALCNTQPCPESWSEWSDWSECEASGVQVRARQCILLFPMGSQCSGNTTESRPCVFDSNFIPEVSVARSSSVEEKRCGE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Growth and Development, Neuronal Cell Markers | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
FLJ12815, sema domain, seven thrombospondin repeats (type 1 and type 1-like), Sema F, SEMAF, semaphorin F, semaphorin-5A, Semaphorin-F, semF, transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5A | |
SEMA5A | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title