Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Semaphorin 5B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25660425UL
Description
Semaphorin 5B Polyclonal specifically detects Semaphorin 5B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Semaphorin 5B | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
FLJ10372, KIAA1445, sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5B | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of human Semaphorin 5B antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
SEMA5B | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QESTLVHPATPNHLHYKGGGTPKNEKYTPMEFKTLNKNNLIPDDRANFYPLQQTNVYTTTYYPSPLNKHSFRPEASPGQRCFPN | |
25 μL | |
Neuroscience | |
54437 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction