Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Semaphorin 6A Antibody (118E7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals DDX0463P100
Description
Semaphorin 6A Monoclonal specifically detects Semaphorin 6A in Human samples. It is validated for Flow Cytometry.Specifications
Semaphorin 6A | |
Monoclonal | |
0.5 mg/mL | |
Flow Cytometry 1:10-1:1000 | |
KIAA1368HT018, SEMA, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A, Sema VIA, SEMA6A1, SEMA6A-1, semaphorin 6A-1, Semaphorin VIA, semaphorin-6A, Semaphorin-6A-1, SEMAQ, VIA | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
1180000000 | |
Unconjugated | |
NP_065847 | |
SEMA6A | |
Sema-6A was used as immunogen. | |
0.1 mg | |
Apoptosis | |
57556 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction