Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Semaphorin 6A Rabbit anti-Human, Clone: 1, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289833100UL
Description
Semaphorin 6A Monoclonal specifically detects Semaphorin 6A in Human samples. It is validated for ELISA.Specifications
Semaphorin 6A | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
1 | |
ELISA 1:5000-1:10000 | |
KIAA1368HT018, SEMA, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A, Sema VIA, SEMA6A1, SEMA6A-1, semaphorin 6A-1, Semaphorin VIA, semaphorin-6A, Semaphorin-6A-1, SEMAQ, VIA | |
Recombinant Human Semaphorin 6A protein | |
100 μg | |
Apoptosis | |
57556 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction