Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Semaphorin 6B Antibody (MM0535-10F7) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
| Antigen | Semaphorin 6B |
|---|---|
| Clone | MM0535-10F7 |
| Concentration | LYOPH |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Semaphorin 6B Monoclonal specifically detects Semaphorin 6B in Human samples. It is validated for Western Blot.Specifications
| Semaphorin 6B | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B, SEMAN, semaphorin VIB, semaphorin Z, semaphorin-6B, semaphorin-6Ba, SEMA-VIB, semaZ, SEM-SEMA-Y, UNQ1907/PRO4353 | |
| SEMA6B | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0535-10F7 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Neuroscience | |
| Q9H3T3 | |
| 10501 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title