Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Semaphorin 6B Antibody (MM0535-10F7), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211880PE
Description
Semaphorin 6B Monoclonal antibody specifically detects Semaphorin 6B in Human samples. It is validated for Western BlotSpecifications
Semaphorin 6B | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
MM0535-10F7 | |
Western Blot | |
sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B, SEMAN, semaphorin VIB, semaphorin Z, semaphorin-6B, semaphorin-6Ba, SEMA-VIB, semaZ, SEM-SEMA-Y, UNQ1907/PRO4353 | |
Human recombinant Semaphorin 6B (extracellular domain) | |
0.1 mL | |
Neuroscience | |
10501 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction