Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Semaphorin 7A Antibody (310829), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB20681H
Description
Semaphorin 7A Monoclonal antibody specifically detects Semaphorin 7A in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| Semaphorin 7A | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Neuroscience | |
| 8482 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 310829 | |
| Flow Cytometry, CyTOF-ready | |
| CD108 antigen, CD108MGC126696, CDw108, H-SEMA-K1, H-Sema-L, JMH, JMH blood group antigen, John-Milton-Hargen human blood group Ag, MGC126692, sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin)7A, sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin)7A (JMH blood group), sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, 7A, Sema K1, Sema L, SEMAK1, SEMALJohn Milton Hagen blood group H-Sema K1, semaphorin 7A, GPI membrane anchor (John Milton Hagen blood group), semaphorin K1, semaphorin L, semaphorin-7A, semaphorin-K1, Semaphorin-L | |
| Mouse myeloma cell line NS0-derived recombinant human Semaphorin 7A, His47-Ala648, Accession # O75326 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction