Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEMG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542098
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542098 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542098 Supplier Invitrogen™ Supplier No. PA542098
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QKGKQQTESK GSFSIQYTYH VDANDHDQSR KSQQYDLNAL HKTTKSQRHL Sequence homology: Human: 100%; Pig: 86%.

The protein encoded by this gene is the predominant protein in semen. The encoded secreted protein is involved in the formation of a gel matrix that encases ejaculated spermatozoa. The prostate-specific antigen (PSA) protease processes this protein into smaller peptides, with each possibly having a separate function. The proteolysis process breaks down the gel matrix and allows the spermatozoa to move more freely.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMG1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SEMG1
Gene Accession No. P04279
Gene Alias Alpha-inhibin-31; Alpha-inhibin-92; BB115391; Cancer/testis antigen 103; CT103; dJ172H20.2; MGC14719; RATSVPIIA; semen coagulating protein; semenoclotin; semenogelin I; Semenogelin-1; SEMG; SEMG1; Seminal basic protein; seminal vesicle protein, secretion 1; seminal vesicle protein, secretion 2; seminal vesicle secretory protein 2; seminal vesicle secretory protein II; SGI; SgI-29; Svp1; Svp-1; Svp5; SVPIIA; SVS II; Svs2; SVS2P; Svs2p2; Svs5; SvsII
Gene Symbols SEMG1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SEMG1 (aa 51-100).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6406
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.