Learn More
Invitrogen™ SEMG1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596341
Description
Immunogen sequence: QKGGSKGRLP SEFSQFPHGQ KGQHYSGQKG KQQTESKGSF SIQYTYHVDA NDHDQSRKSQ QYDLNALHKT TKSQRHLGGS QQLLHNKQEG RDHDKSKGHF HRVVIHHKGG KAHRGTQNPS QDQGNSPSGK GISSQYSNTE ERLWVHGLSK EQTSVSGAQK GRKQGGSQSS YVLQTEELVA NKQQRETKNS HQNKGHYQNV VEVREEHSSK VQTSLCPAHQ DKLQHGSKDI FSTQDELLVY NKNQHQTKNL NQDQQHGRKA NKISYQSSST EERRLHYGEN GVQKDVS; Positive Samples: THP-1, HT-1080.
The protein encoded by this gene is the predominant protein in semen. The encoded secreted protein is involved in the formation of a gel matrix that encases ejaculated spermatozoa. The prostate-specific antigen (PSA) protease processes this protein into smaller peptides, with each possibly having a separate function. The proteolysis process breaks down the gel matrix and allows the spermatozoa to move more freely.
Specifications
SEMG1 | |
Polyclonal | |
Unconjugated | |
SEMG1 | |
Alpha-inhibin-31; Alpha-inhibin-92; BB115391; Cancer/testis antigen 103; CT103; dJ172H20.2; MGC14719; RATSVPIIA; semen coagulating protein; semenoclotin; semenogelin I; Semenogelin-1; SEMG; SEMG1; Seminal basic protein; seminal vesicle protein, secretion 1; seminal vesicle protein, secretion 2; seminal vesicle secretory protein 2; seminal vesicle secretory protein II; SGI; SgI-29; Svp1; Svp-1; Svp5; SVPIIA; SVS II; Svs2; SVS2P; Svs2p2; Svs5; SvsII | |
Rabbit | |
Affinity Chromatography | |
RUO | |
25013, 53878, 6406 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.79 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04279 | |
SEMG1, Svs2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 24-310 of human SEMG1 (NP_0029981). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.