Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEMG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560109
Description
Immunogen sequence: EERHLNCGEK GIQKGVSKGS ISIQTEEQIH GKSQNQVRIP SQAQEYGHKE Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 33%.
The secreted protein encoded by this gene is involved in the formation of a gel matrix that encases ejaculated spermatozoa. Proteolysis by the prostate-specific antigen (PSA) breaks down the gel matrix and allows the spermatozoa to move more freely. The encoded protein is found in lesser abundance than a similar semenogelin protein. The genes encoding these two semenogelin proteins are found in a cluster on chromosome 20.
Specifications
SEMG2 | |
Polyclonal | |
Unconjugated | |
SEMG2 | |
9530026M05Rik; BB121242; Semenogelin 2; semenogelin II; Semenogelin-2; Semg2; seminal vesicle protein 3; seminal vesicle secretion 3; seminal vesicle secretion 3 alpha; seminal vesicle secretion 3A; seminal vesicle secretory protein 3A; Seminal vesicle secretory protein III; SGII; Svp3; Svp-3; SVS III; Svs3; Svs3a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6407 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q02383 | |
SEMG2 | |
Recombinant protein corresponding to Human SEMG2. Recombinant protein control fragment (Product #RP-99340). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction