Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEMG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588785
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588785 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588785 Supplier Invitrogen™ Supplier No. PA588785
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HGKSQNQVRI PSQAQEYGHK ENKISYQSSS TEERRLNSGE KDVQKGVSKG SISIQTEEKI HGKSQNQVTI PSQDQEHGHK ENKMSYQSSS TEERRLNYGG KSTQKDVSQS SISFQIEKLV EGKSQIQTPN PNQDQWSGQN AKGKSGQSAD SKQDLLSHEQ KGRYKQESSE SHNIVITEHE VAQDDHLTQQ YNEDRNPIST; Positive Samples: 293T, HeLa, Mouse testis.

The secreted protein encoded by this gene is involved in the formation of a gel matrix that encases ejaculated spermatozoa. Proteolysis by the prostate-specific antigen (PSA) breaks down the gel matrix and allows the spermatozoa to move more freely. The encoded protein is found in lesser abundance than a similar semenogelin protein. The genes encoding these two semenogelin proteins are found in a cluster on chromosome 20.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMG2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SEMG2
Gene Accession No. Q02383
Gene Alias 9530026M05Rik; BB121242; Semenogelin 2; semenogelin II; Semenogelin-2; Semg2; seminal vesicle protein 3; seminal vesicle secretion 3; seminal vesicle secretion 3 alpha; seminal vesicle secretion 3A; seminal vesicle secretory protein 3A; Seminal vesicle secretory protein III; SGII; Svp3; Svp-3; SVS III; Svs3; Svs3a
Gene Symbols SEMG2, Svs3a
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 383-582 of human SEMG2 (NP_0029991).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6407, 64335
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.