Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semiquantitative Standard 1, Specpure™

Catalog No. p-7080279
Click to view available options
Quantity:
100 mL
500 mL

Catalog No. AA47180AE

Supplier: Thermo Scientific Chemicals 047180AE

Only null left
Add to Cart

Semiquantitative Standard 1, Specpure™ is used as a standard for inductively coupled plasma and direct current plasma (DCP) emission spectrometry for the analysis of trace elements in biomedical and environmental samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Semiquantitative Standard 1
Physical Form Liquid
MDL Number MFCD02091742
UN Number UN3093
Solubility Miscible with water
Quantity 100 mL
Grade Specpure

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.