Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SEN 177
GREENER_CHOICE

Catalog No. 697910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-791-0 10 mg
69-795-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 69-791-0 Supplier Tocris Bioscience™ Supplier No. 6979/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent glutaminyl cyclase inhibitor; disrupts binding of SIRP-α to CD47

SEN 177 is a potent glutaminyl cyclase (QPCT) inhibitor (IC50 = 53 nM). SEN 177 reduces aggregation of mutant huntingtin (mHTT) through inhibition of QPCT and reduces Q80 aggregates in vitro. Inhibition of QPCT also reduces aggregate formation in in vivo models of Huntington's disease. SEN 177 potently inhibits QPCT-like protein (IC50 = 13 nM) and disrupts binding of SIRP-α to CD47. The compound increases antibody-dependent cellular phagocytosis and cytotoxicity in cancer cell lines.

Specifications

Chemical Name or Material 6 '-Fluoro-2-[4-(4-methyl-4 H-1,2,4-triazol-3-yl)-1-piperidinyl]-3,3 '-bipyridine
CAS 2117405-13-5
Quantity 10 mg
Target Other Transferase Inhibitors
Molecular Formula C18H19FN6
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.