Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Senataxin Antibody (4G1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280943
Description
Senataxin Monoclonal specifically detects Senataxin in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready.Specifications
Senataxin | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry : 5 μg/mL, Immunohistochemistry 1:200-1:500, Immunohistochemistry-Paraffin : 1:200-1:500, Flow (Intracellular) : 5 μg/mL, CyTOF-ready | |
ALS4recessive, non-Friedreich type 1, AOA2FLJ43459, DKFZp781B151, KIAA0625FLJ12840, senataxin, SETX | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
4G1 | |
Unconjugated | |
PBS | |
SETX | |
Partial recombinant human Senataxin protein (between amino acids 2300-2600) [UniProt# Q7Z333] | |
0.1 mg | |
DNA Repair | |
23064 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction