Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Senataxin Antibody (5D8), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP252727AF532
Description
Senataxin Monoclonal specifically detects Senataxin in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
Senataxin | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
SETX | |
Partial recombinant human Senataxin protein (between amino acids 2300-2600) [UniProt Q7Z333] | |
Primary | |
Human |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
5D8 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular) | |
ALS4recessive, non-Friedreich type 1, AOA2FLJ43459, DKFZp781B151, KIAA0625FLJ12840, senataxin, SETX | |
Mouse | |
0.1 mL | |
23064 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction