Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Senataxin Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10057543AF350
Description
Senataxin Polyclonal specifically detects Senataxin in Human samples. It is validated for Western Blot, Knockout Validated, Knockdown Validated.Specifications
Senataxin | |
Polyclonal | |
Western Blot, Knockout Validated, KnockDown Validated | |
ALS4recessive, non-Friedreich type 1, AOA2FLJ43459, DKFZp781B151, KIAA0625FLJ12840, senataxin, SETX | |
Rabbit | |
100 μL | |
23064 | |
Store at 4°C in the dark. |
Western Blot, Immunoassay, KnockDown | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
SETX | |
The immunogen recognized by this antibody maps to a region between residue 2627 and 2677 of human senataxin (NP_055861.2). | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction