Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Senataxin Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10057543AF488
Description
Senataxin Polyclonal specifically detects Senataxin in Human samples. It is validated for Western Blot, Knockout Validated, Knockdown Validated.Specifications
Senataxin | |
Polyclonal | |
Western Blot, Knockout Validated, KnockDown Validated | |
ALS4recessive, non-Friedreich type 1, AOA2FLJ43459, DKFZp781B151, KIAA0625FLJ12840, senataxin, SETX | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, Immunoassay, KnockDown | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SETX | |
The immunogen recognized by this antibody maps to a region between residue 2627 and 2677 of human senataxin (NP_055861.2). | |
100 μL | |
DNA Repair | |
23064 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction