Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Senataxin Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10057542B
Description
Senataxin Polyclonal specifically detects Senataxin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
Senataxin | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated | |
ALS4recessive, non-Friedreich type 1, AOA2FLJ43459, DKFZp781B151, KIAA0625FLJ12840, senataxin, SETX | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
Biotin | |
PBS with 0.05% Sodium Azide | |
SETX | |
The immunogen recognized by this antibody maps to a region between residue 875 and 925 of human senataxin using the numbering given in entry NP_055861.2 (GeneID 23064). | |
100 μL | |
DNA Repair | |
23064 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction