Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sennoside A, ≥96% (HPLC), MilliporeSigma™ Supelco™

Catalog No. p-7187494 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5606 5 mg
11-101-5607 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5606 Supplier MilliporeSigma Supelco Supplier No. 689095MGF
Only null left
Add to Cart
Add to Cart

Sennoside A is a hydroxy anthracene glycoside generally present in the polyherbal formulations that contain Cassia angustifolia leaves, which is used in the treatment of constipation.

Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 81-27-6
Physical Form Neat
Linear Formula C42H38O20
MDL Number MFCD00151527
Type Analytical Standard
Quantity 5 mg
Formula Weight 862.74
Percent Purity ≥96% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.