Learn More
SENP1 Mouse anti-Human, Clone: OTI1D7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800001

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SENP1 is a protease that catalyzes two essential functions in the SUMO pathway: processing of full-length SUMO1, SUMO2 and SUMO3 to their mature forms and deconjugation of SUMO1, SUMO2 and SUMO3 from targeted proteins. SENP deconjugates SUMO1 from HIPK2 and from HDAC1, which decreases the transcriptional repression activity of the latter.Specifications
SENP1 | |
Monoclonal | |
Unconjugated | |
SENP1 | |
SuPr-2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
29843 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI1D7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9P0U3 | |
SENP1 | |
Human recombit protein fragment corresponding to amino acids 221-460 of human SENP1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.