Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SENP1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA588971

Catalog No. PIPA588971


Only null left
Add to Cart

Description

Description

Immunogen sequence: SRCLSSSKNT LKDSLFKNGN SCASQIIGSD TSSSGSASIL TNQEQLSHSV YSLSSYTPDV AFGSKDSGTL HHPHHHHSVP HQPDNLAASN TQSEGSDSVI LLKVKDSQTP TPSSTFFQAE LWIKELTSVY DSRARERLRQ IEEQKALALQ LQNQRLQERE HSVHDSVELH LRVPLEKEIP VTVVQETQKK GHKLTDSEDE F; Positive Samples: BxPC-3, HT-29, HepG2, HeLa, Mouse testis, Mouse thymus, Rat testis; Cellular Location: Cytoplasm, Nucleus.

SENP1 is a protease that catalyzes two essential functions in the SUMO pathway: processing of full-length SUMO1, SUMO2 and SUMO3 to their mature forms and deconjugation of SUMO1, SUMO2 and SUMO3 from targeted proteins. SENP deconjugates SUMO1 from HIPK2 and from HDAC1, which decreases the transcriptional repression activity of the latter.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SENP1
Polyclonal
Unconjugated
SENP1
2310046A20Rik; D15Ertd528e; E330036L07Rik; SEN1; Senp1; sentrin/SUMO-specific protease SENP1; Sentrin-specific protease 1; SUMO-1 protease 2; SUMO1/sentrin specific peptidase 1; SUMO1/sentrin specific protease 1; Sumo1/sentrin/SMT3 specific peptidase 1; Supr2; suPr-2
Rabbit
Affinity Chromatography
RUO
223870, 29843, 300193
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
2.32 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
P59110, Q9P0U3
SENP1
Recombinant fusion protein containing a sequence corresponding to amino acids 220-420 of human SENP1 (NP_0012545231).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.