Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SENP1 Recombinant Rabbit Monoclonal Antibody (JG37-79)
GREENER_CHOICE

Catalog No. PIMA534767
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534767 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534767 Supplier Invitrogen™ Supplier No. MA534767
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: K562, LOVO, SH-SY-5Y, SiHa, Daudi, human liver tissue, human small Intestine tissue.

SENP1 is a protease that catalyzes two essential functions in the SUMO pathway: processing of full-length SUMO1, SUMO2 and SUMO3 to their mature forms and deconjugation of SUMO1, SUMO2 and SUMO3 from targeted proteins. SENP deconjugates SUMO1 from HIPK2 and from HDAC1, which decreases the transcriptional repression activity of the latter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SENP1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JG37-79
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene SENP1
Gene Accession No. Q9P0U3
Gene Alias 2310046A20Rik; D15Ertd528e; E330036L07Rik; SEN1; Senp1; sentrin/SUMO-specific protease SENP1; Sentrin-specific protease 1; SUMO-1 protease 2; SUMO1/sentrin specific peptidase 1; SUMO1/sentrin specific protease 1; Sumo1/sentrin/SMT3 specific peptidase 1; Supr2; suPr-2
Gene Symbols SENP1
Host Species Rabbit
Immunogen Recombinant protein within Human SENP1 aa 380-521.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29843, 300193
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.