Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SENP2 Antibody (OTI3D3), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274062APC
Description
SENP2 Monoclonal specifically detects SENP2 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Flow Cytometry.Specifications
| SENP2 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| SENP2 | |
| Recombinant protein fragment corresponding to amino acids 139-523 of human SENP2 (NP_067640) produced in E.coli. | |
| 0.1 mL | |
| 59343.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI3D3 | |
| Flow Cytometry | |
| AXAM2Axam2, DKFZp762A2316, EC 3.4.22, EC 3.4.22.68, KIAA1331sentrin-specific protease 2, Sentrin/SUMO-specific protease SENP2, Smt3ip2, SMT3IP2EC 3.4.22.-, SMT3-specific isopeptidase 2, SUMO1/sentrin/SMT3 specific peptidase 2, SUMO1/sentrin/SMT3 specific protease 2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Monkey, Primate | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction