Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SENP2 Monoclonal Antibody (OTI3H10), TrueMAB™, OriGene
SDP

Catalog No. 502540067
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0067 30 μL
50-167-5464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0067 Supplier OriGene Supplier No. TA504864S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SENP2 Monoclonal antibody specifically detects SENP2 in Human samples. It is validated for Western Blot

Specifications

Antigen SENP2
Applications Western Blot
Classification Monoclonal
Clone OTI3H10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Acbd3
Gene Accession No. Q9H3P7
Gene Alias 2310007L05Rik; 4930538C18Rik; AI646780; AW554757; Axam; Axam2; Axin-associating molecule; CT; EGK_11819; KIAA1331; mKIAA1331; Senp2; sentrin/SUMO-specific protease SENP2; Sentrin-specific protease 2; Smt3ip2; SMT3-specific isopeptidase 2; SUMO specific peptidase 2; SUMO/sentrin specific peptidase 2; SUMO/sentrin specific protease 2; SUMO-1 protease 1; SUMO-1 protease-1; SUMO1/sentrin/SMT3 specific peptidase 2; SUMO1/sentrin/SMT3 specific protease 2; SUMO-1/Smt3-specific isopeptidase 2; Supr1; SuPr-1
Host Species Mouse
Immunogen Full length human recombinant protein of human ACBD3 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64746
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.