Learn More
SENP2 Mouse anti-Human, Clone: OTI1B8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504871

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SUMO is a small ubiquitin-like protein that can be covalently conjugated to other proteins. SENP2 is one of a group of enzymes that process newly synthesized SUMO1, SUMO2, and SUMO3 into the conjugatable mature forms and catalyze the deconjugation of these same SUMO proteins from their targeted substrates. SENP2 may also down-regulate CTNNB1 levels and thereby modulate the Wnt pathway.Specifications
SENP2 | |
Monoclonal | |
Unconjugated | |
SENP2 | |
AXAM2, SMT3IP2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
59343 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1B8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9HC62 | |
SENP2 | |
Human recombit protein fragment corresponding to amino acids 139-523 of human SENP2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.