Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SENP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120603
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120603 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120603 Supplier Invitrogen™ Supplier No. PA5120603
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: rat lung, rat testis. Immunogen sequence: LELTEDMEKE ISNALGHGPQ DEILSSAFKL RITRGDIQTL KNYHWLNDEV INFYMNLLVE RNKKQGYPAL HVFSTFFYPK LKSGGYQAVK RWTKGVNLFE QEIILVPIHR KVHWSLVVID LRKKCLKYLD SMGQKGHRIC EILLQYLQDE SKTKRNSDLN LLEWTHHSMK PHEIPQQLNG SDCGMFTCKY ADYISRDKPI TFTQHQMPLF RKKMVWEILH QQLL.

SUMO is a small ubiquitin-like protein that can be covalently conjugated to other proteins. SENP2 is one of a group of enzymes that process newly synthesized SUMO1, SUMO2, and SUMO3 into the conjugatable mature forms and catalyze the deconjugation of these same SUMO proteins from their targeted substrates. SENP2 may also down-regulate CTNNB1 levels and thereby modulate the Wnt pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SENP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SENP2
Gene Accession No. Q91ZX6, Q9EQE1
Gene Alias 2310007L05Rik; 4930538C18Rik; AI646780; AW554757; Axam; Axam2; Axin-associating molecule; CT; EGK_11819; KIAA1331; mKIAA1331; Senp2; sentrin/SUMO-specific protease SENP2; Sentrin-specific protease 2; Smt3ip2; SMT3-specific isopeptidase 2; SUMO specific peptidase 2; SUMO/sentrin specific peptidase 2; SUMO/sentrin specific protease 2; SUMO-1 protease 1; SUMO-1 protease-1; SUMO1/sentrin/SMT3 specific peptidase 2; SUMO1/sentrin/SMT3 specific protease 2; SUMO-1/Smt3-specific isopeptidase 2; Supr1; SuPr-1
Gene Symbols SENP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 366-589 of human SENP2 (NP_067640.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 75826, 78973
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.