Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SENP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584450
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584450 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584450 Supplier Invitrogen™ Supplier No. PA584450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LREEHVTCVQ SILDEFLQTY GSLIPLSTDE VVEKLEDIFQ QEFSTPSRKG LVLQLIQSYQ RMPGNAMVRG FRVAYKRHVL TMDDLGTL Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 98%.

The reversible posttranslational modification of proteins by the addition of small ubiquitin-like SUMO proteins is required for numerous biologic processes. SUMO-specific proteases, such as SENP3, are responsible for the initial processing of SUMO precursors to generate a C-terminal diglycine motif required for the conjugation reaction. They also have isopeptidase activity for the removal of SUMO from high molecular mass SUMO conjugates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SENP3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Senp3
Gene Accession No. Q9H4L4
Gene Alias AA408656; DKFZP586K0919; DKFZp762A152; SENP3; sentrin/SUMO-specific protease 3; sentrin/SUMO-specific protease SENP3; Sentrin-specific protease 3; Smt3ip; Smt3ip1; Smt3-specific isopeptidase 1; SSP3; SUMO specific peptidase 3; SUMO/sentrin specific peptidase 3; SUMO/sentrin specific protease 3; SUMO1/sentrin/SMT3 specific peptidase 3; SUMO1/sentrin/SMT3 specific protease 3; SUMO-1-specific protease 3; SUSP3; Ulp1
Gene Symbols Senp3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SENP3. Recombinant protein control fragment (Product #RP-101397).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26168
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.