Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SENP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675593
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-593 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-593 Supplier Invitrogen™ Supplier No. PA141239
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: HeLa or NIH-3T3 whole cell lysate. Immunogen sequence: CNRKPFINRE ITNYRARH.

SUMO (small ubiquitin-related modifier), a member of the ubiquitin-like protein family, regulates diverse cellular functions of a variety of target proteins including transcription, DNA repair, nucleocytoplasmic trafficking and chromosome segregation. SUMO precursor proteins undergo cleavage of the residues after the 'GG' region by SUMO-specific proteases in maturation. This cleavage of the precursor is a prerequisite for subsequent sumoylation. The sentrin-specific (or SUMO-specific) protease (SENP) proteins belong to the peptidase C48 family and include SENP1-3 and SENP5-8. SENP1, SENP2 and SENP3 degrade UBL1 and SMT3H2 conjugates and subsequently release the monomers from sumoylated substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SENP5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene SENP5
Gene Accession No. Q6NXL6, Q96HI0
Gene Alias 6230429P13Rik; A730063F07Rik; AI851888; BB189556; DKFZp564O1016; FKSG45; FLJ42398; MGC27076; Senp5; sentrin/SUMO-specific protease SENP5; Sentrin-specific protease 5; SMT3IP3; SUMO/sentrin specific peptidase 5; SUMO/sentrin specific protease 5; SUMO/Smt3-specific isopeptidase 3; SUMO1/sentrin specific peptidase 5; SUMO1/sentrin specific protease 5; Sumo1/sentrin/SMT3 specific peptidase 5
Gene Symbols SENP5
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 229-246 of human SENP5.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 205564, 320213
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.