Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SENP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575186
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575186 Supplier Invitrogen™ Supplier No. PA575186
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Ubiquitin-like molecules, such as SUMO1, are structurally related to ubiquitin. and can be ligated to target proteins in a similar manner as ubiquitin. However, covalent attachment of UBLs does not result in degradation of the modified proteins. SUMO1 modification is implicated in the targeting of RANGAP1. to the nuclear pore complex, as well as in stabilization of I-kappa-B-alpha from degradation by the 26S proteasome. Like ubiquitin, UBLs are synthesized as precursor proteins, with 1 or more amino acids following the C-terminal glycine-glycine residues of the mature UBL protein. Thus, the tail sequences of the UBL precursors need to be removed by UBL-specific proteases, such as SENP6, prior to their conjugation to target proteins. SENPs also display isopeptidase activity for deconjugation of SUMO-conjugated substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SENP6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene SENP6
Gene Accession No. Q9GZR1
Gene Alias 2810017C20Rik; E130319N12Rik; FKSG6; FLJ11355; FLJ11887; KIAA0389; KIAA0797; mKIAA0797; RP1-134M13.1; SENP6; sentrin/SUMO-specific protease SENP6; Sentrin-specific protease 6; Sentrin-specific protease 6-like protein; SSP1; SUMO specific peptidase 6; SUMO/sentrin specific peptidase 6; SUMO/sentrin specific protease 6; SUMO1/sentrin specific peptidase 6; SUMO1/sentrin specific protease 6; Sumo1/sentrin/SMT3 specific peptidase 6; SUMO-1-specific protease 1; Susp1
Gene Symbols SENP6
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 1040-1089 of Human SENP6.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26054, 300860
Target Species Human, Rat, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.