Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SENP7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582961
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582961 Supplier Invitrogen™ Supplier No. PA582961
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STISTEFEKP SENYHQDPKL PEEITTKPTK SDFTKLSSLN SQELTLSNAT KSASAGSTTE TVENSNSIDI VGISSLVEKD ENELNTIEKP ILRGHNEGNQ SLISAEPIVV SSDEEGPVEH KSSEILKLQS KQDRE Highest antigen sequence indentity to the following orthologs: Rat - 47%, Mouse - 41%.

SENP7 is a protease that may be involved in two essential functions in the SUMO pathway: processing of full-length SUMO1, SUMO2 and SUMO3 to their mature conjugatable forms and/or deconjugation of SUMO1, SUMO2 and SUMO3 from targeted substrate proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SENP7
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SENP7
Gene Accession No. Q9BQF6
Gene Alias 2410152H17Rik; 2810413I22Rik; 2900036C23Rik; 6030449K19Rik; AI790676; KIAA1707; SENP7; sentrin/SUMO-specific protease SENP7; Sentrin-specific protease 7; SSP2; SUMO1/sentrin specific peptidase 7; SUMO1/sentrin specific protease 7; Sumo1/sentrin/SMT3 specific peptidase 7; SUMO-1-specific protease 2; Susp2
Gene Symbols SENP7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SENP7. Recombinant protein control fragment (Product #RP-89269).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57337
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.