Learn More
Invitrogen™ SENP8 Monoclonal Antibody (OTI3E4)

Description
SENP8 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | SENP8 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI3E4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Senp8 |
| Gene Accession No. | Q96LD8 |
| Gene Alias | 9130010J17Rik; AU020827; DEN1; deneddylase 1; deneddylase-1; FKSG8; NEDD8 specific-protease cysteine 2; NEDD8-specific protease 1; Nedp1; protease, cysteine 2; protease, cysteine, 2 (NEDD8 specific); PRSC2; SENP8; sentrin/SUMO-specific protease SENP8; Sentrin-specific protease 8; SUMO sentrin specific protease family member 8; SUMO/sentrin peptidase family member, NEDD8 specific; SUMO/sentrin specific peptidase 8; SUMO/sentrin specific peptidase family member 8; SUMO/sentrin specific protease 8; SUMO/sentrin specific protease family member 8; Sumo1/sentrin/SMT3 specific peptidase 8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.