Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SENP8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118500
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118500 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118500 Supplier Invitrogen™ Supplier No. PA5118500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SENP8.

SENP8 (Cysteine protease FKSG8; DEN1; FKSG8; HsT17512; NEDP1; PRSC2; SUMO/sentrin specific protease family member 8), is a novel cysteine protease which belongs to the ULP family of deubiquitinases. ULP family members consist of a C-terminal core catalytic domain and N-terminal extensions determining substrate specificity. SENP8, a 24KDa protein, plays a major role in regulating NEDD8 pathway. It processes preNEDD8 to its mature form and deconjugates NEDD8 from substrates such as Cullins and p53. SENP8 may play a role in preventing accumulation of hyper-neddylated cullin and maintaining the level of mono-neddylated cullin that is thought to increase the efficiency of ubiquitination by SCF ubiquitin ligases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SENP8
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Senp8
Gene Accession No. Q5FVJ8, Q96LD8, Q9D2Z4
Gene Alias 9130010J17Rik; AU020827; DEN1; deneddylase 1; deneddylase-1; FKSG8; NEDD8 specific-protease cysteine 2; NEDD8-specific protease 1; Nedp1; protease, cysteine 2; protease, cysteine, 2 (NEDD8 specific); PRSC2; SENP8; sentrin/SUMO-specific protease SENP8; Sentrin-specific protease 8; SUMO sentrin specific protease family member 8; SUMO/sentrin peptidase family member, NEDD8 specific; SUMO/sentrin specific peptidase 8; SUMO/sentrin specific peptidase family member 8; SUMO/sentrin specific protease 8; SUMO/sentrin specific protease family member 8; Sumo1/sentrin/SMT3 specific peptidase 8
Gene Symbols Senp8
Host Species Rabbit
Immunogen A synthesized peptide derived from human SENP8(Accession Q96LD8), corresponding to amino acid residues T171-K212.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123228, 315723, 71599
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.