Learn More
Sensidyne™ Colorimetric Gas Detector Tubes: Cyclohexanone
SDP
Supplier:  Sensidyne™ 197U
Catalog No. 18-068T
Only null left
Add to Cart
Description
Description
Specifications
Specifications
Cyclohexanone
10 Tubes/Box
0°C to 40°C
2 to 100ppm
32°F to 104°F
Standard tube
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.