Learn More
Sensidyne™ Colorimetric Gas Detector Tubes: Vinyl Chloride
SDP
Catalog No. 18074E Shop All Sensidyne Inc. Products
Click to view available options
Measurement Range:
5 to 500ppm
Catalog No. Measurement Range
18-074E 5 to 500ppm
1 options
Catalog No. 18-074E Supplier Sensidyne™ Supplier No. 132SB
Only null left
Add to Cart
Add to Cart
Specifications
Components Measured Vinyl Chloride
Measurement Range 5 to 500ppm
Quantity 2x5 tubes/box
Tube Type Standard tube