Description
- Easy to use detector tubes
- Provides low-cost method of routine or spot sampling for gases and vapors
- Sold in packs of 10
Specifications
Specifications
| Components Measured | Carbon Dioxide |
| Measurement Range | 0.01 to 0.70% |
| No. of Strokes | n/a |
| Quantity | 10 |
| Response Time | Not Applicable |