Learn More
Sensidyne™ Detector Tubes: Carbon Disulfide
Detector tubes
Supplier: Sensidyne™ 141SA
Description
All detector tubes are sold in packs of 10.Specifications
Carbon Disulfide | |
1 | |
2 min. | |
0°C to 40°C |
30 to 500 | |
10 | |
32°F to 104°F |