Learn More
Sensidyne™ Detector Tubes: Hydrogen Sulfide
SDP
Supplier:  Sensidyne™ 120UH
 View more versions of this product
Catalog No. 18-070Q
Only null left
Add to Cart
Description
Description
Detector tubes are sold in packs of 10
Specifications
Specifications
Hydrogen Sulfide
1.5
1 min.
0°C to 40°C
2 to 20% vol.
10
32°F to 104°F
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.