Learn More
Sensidyne™ Detector Tubes: Propane
SDP
Supplier:  Sensidyne™ 125SA
Catalog No. 18-073F
Only null left
Add to Cart
Description
Description
All detector tubes are sold in packs of 10.
Specifications
Specifications
Propane
1
4 min.
0°C to 40°C
0.02 to 0.5% vol.
10
32°F to 104°F
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.